missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3,4-dicarboximide

Catalog No. AAH3175806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3175806 5 g
AAH3175814 25 g
AAH3175822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3175806 Supplier Thermo Scientific Chemicals Supplier No. H3175806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4664-01-1 | C7H4N2O2 | 148.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4664-01-1
MDL Number MFCD00013439

Specifications

Melting Point 230°C to 232°C
Molecular Formula C7H4N2O2
Quantity 5 g
Synonym 3,4-pyridinedicarboximide, cinchomeronimide, 3,4-pyridinecarboximide, 1h-pyrrolo 3,4-c pyridine-1,3 2h-dione, 2h-pyrrolo 3,4-c pyridine-1,3-dione, pyrrolo 3,4-c pyridine-1,3-dione, cinchomeronic acid imide, 4-azaphthalimid, pyridine-3,4-dicarboximide, 1h,2h,3h-pyrrolo 3,4-c pyridine-1,3-dione
InChI Key SJSABZBUTDSWMJ-UHFFFAOYSA-N
SMILES O=C1NC(=O)C2=CN=CC=C12
IUPAC Name pyrrolo[3,4-c]pyridine-1,3-dione
Molecular Weight (g/mol) 148.12
PubChem CID 72926
Formula Weight 148.12
Chemical Name or Material Pyridine-3,4-dicarboximide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber US7966000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only