Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyridine-2-sulfonic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 252°C |
| Color | White-Yellow |
| Quantity | 5 g |
| UN Number | 3261 |
| Formula Weight | 159.16 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pyridine-2-sulfonic Acid |
Safety and Handling
| TSCA | No |