missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine-2-carboxylic Acid Hydrochloride 98.0+%, TCI America™
$124.89 - $397.05
Chemical Identifiers
| CAS | 636-80-6 |
|---|---|
| Molecular Formula | C6H6ClNO2 |
| Molecular Weight (g/mol) | 159.57 |
| MDL Number | MFCD00050666 |
| InChI Key | HFAFXVOPGDBAOK-UHFFFAOYSA-N |
| Synonym | Picolinic Acid Hydrochloride |
| PubChem CID | 71559 |
| IUPAC Name | hydrogen pyridine-2-carboxylic acid chloride |
| SMILES | [H+].[Cl-].OC(=O)C1=CC=CC=N1 |
Chemical Identifiers
| 636-80-6 | |
| 159.57 | |
| HFAFXVOPGDBAOK-UHFFFAOYSA-N | |
| 71559 | |
| [H+].[Cl-].OC(=O)C1=CC=CC=N1 |
| C6H6ClNO2 | |
| MFCD00050666 | |
| Picolinic Acid Hydrochloride | |
| hydrogen pyridine-2-carboxylic acid chloride |
Specifications
| 636-80-6 | |
| C6H6ClNO2 | |
| 5 g | |
| HFAFXVOPGDBAOK-UHFFFAOYSA-N | |
| hydrogen pyridine-2-carboxylic acid chloride | |
| 71559 | |
| ≥98.0% (HPLC,T) | |
| Pyridine-2-carboxylic Acid Hydrochloride |
| White | |
| MFCD00050666 | |
| Picolinic Acid Hydrochloride | |
| [H+].[Cl-].OC(=O)C1=CC=CC=N1 | |
| 159.57 | |
| 159.57 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes