missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-carboxylic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. P04225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P04225G 5 g
P042225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P04225G Supplier TCI America Supplier No. P04225G
Only null left

Chemical Identifiers

CAS 636-80-6
Molecular Formula C6H6ClNO2
Molecular Weight (g/mol) 159.57
MDL Number MFCD00050666
InChI Key HFAFXVOPGDBAOK-UHFFFAOYSA-N
Synonym Picolinic Acid Hydrochloride
PubChem CID 71559
IUPAC Name hydrogen pyridine-2-carboxylic acid chloride
SMILES [H+].[Cl-].OC(=O)C1=CC=CC=N1

Specifications

Color White
Quantity 5 g
Formula Weight 159.57
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-2-carboxylic Acid Hydrochloride