Learn More
Pyridine-2-carboxamidoxime, 97%
CAS: 1772-01-6 | C6H7N3O | 137.142 g/mol
$128.86 - $432.49
Chemical Identifiers
| CAS | 1772-01-6 |
|---|---|
| Molecular Formula | C6H7N3O |
| Molecular Weight (g/mol) | 137.142 |
| MDL Number | MFCD00085159 |
| InChI Key | NJNUYQDYGLUAAG-WAYWQWQTSA-N |
| Synonym | n'-hydroxypyridine-2-carboximidamide, pyridine-2-carboxamide oxime, 2-pyridylamide oxime, z-n'-hydroxypicolinimidamide, z-n'-hydroxypyridine-2-carboximidamide, pyridin-2-amidoxim, 2-pyridylamidoxime, z-n'-hydroxypicolinimidam ide, 2-pyridinecarboximidamide, n'-hydroxy, z-nitroso 1h-pyridin-2-ylidene methanamine |
| PubChem CID | 5712435 |
| IUPAC Name | (Z)-nitroso(1H-pyridin-2-ylidene)methanamine |
| SMILES | C1=CC(=C(N)N=O)NC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5170503
|
Thermo Scientific Chemicals
H5170503 |
1 g |
Each for $128.86
|
|
|||||
|
AAH5170506
|
Thermo Scientific Chemicals
H5170506 |
5 g |
Each for $432.49
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1772-01-6 | |
| 137.142 | |
| NJNUYQDYGLUAAG-WAYWQWQTSA-N | |
| 5712435 | |
| C1=CC(=C(N)N=O)NC=C1 |
| C6H7N3O | |
| MFCD00085159 | |
| n'-hydroxypyridine-2-carboximidamide, pyridine-2-carboxamide oxime, 2-pyridylamide oxime, z-n'-hydroxypicolinimidamide, z-n'-hydroxypyridine-2-carboximidamide, pyridin-2-amidoxim, 2-pyridylamidoxime, z-n'-hydroxypicolinimidam ide, 2-pyridinecarboximidamide, n'-hydroxy, z-nitroso 1h-pyridin-2-ylidene methanamine | |
| (Z)-nitroso(1H-pyridin-2-ylidene)methanamine |
Specifications
| 1772-01-6 | |
| C6H7N3O | |
| 1 g | |
| NJNUYQDYGLUAAG-WAYWQWQTSA-N | |
| (Z)-nitroso(1H-pyridin-2-ylidene)methanamine | |
| 5712435 | |
| 97% |
| 115°C to 120°C | |
| MFCD00085159 | |
| n'-hydroxypyridine-2-carboximidamide, pyridine-2-carboxamide oxime, 2-pyridylamide oxime, z-n'-hydroxypicolinimidamide, z-n'-hydroxypyridine-2-carboximidamide, pyridin-2-amidoxim, 2-pyridylamidoxime, z-n'-hydroxypicolinimidam ide, 2-pyridinecarboximidamide, n'-hydroxy, z-nitroso 1h-pyridin-2-ylidene methanamine | |
| C1=CC(=C(N)N=O)NC=C1 | |
| 137.142 | |
| 137.14 | |
| Pyridine-2-carboxamidoxime |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
RTECSNumber : TJ4310000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only