missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-carboxamidoxime, 97%

Catalog No. AAH5170506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5170506 5 g
AAH5170503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5170506 Supplier Thermo Scientific Chemicals Supplier No. H5170506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1772-01-6 | C6H7N3O | 137.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1772-01-6
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.142
MDL Number MFCD00085159
InChI Key NJNUYQDYGLUAAG-WAYWQWQTSA-N
Synonym n'-hydroxypyridine-2-carboximidamide, pyridine-2-carboxamide oxime, 2-pyridylamide oxime, z-n'-hydroxypicolinimidamide, z-n'-hydroxypyridine-2-carboximidamide, pyridin-2-amidoxim, 2-pyridylamidoxime, z-n'-hydroxypicolinimidam ide, 2-pyridinecarboximidamide, n'-hydroxy, z-nitroso 1h-pyridin-2-ylidene methanamine
PubChem CID 5712435
IUPAC Name (Z)-nitroso(1H-pyridin-2-ylidene)methanamine
SMILES C1=CC(=C(N)N=O)NC=C1

Specifications

Melting Point 115°C to 120°C
Quantity 5 g
Formula Weight 137.14
Percent Purity 97%
Chemical Name or Material Pyridine-2-carboxamidoxime
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber TJ4310000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only