missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-boronic acid dimethyl ester, 95%, Thermo Scientific™

Catalog No. p-7035350
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53067MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH53067MD Supplier Thermo Scientific Chemicals Supplier No. H53067MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 136805-54-4
Molecular Formula C7H10BNO2
Molecular Weight (g/mol) 150.972
MDL Number MFCD03412109
InChI Key ITSIDKYMJQVPQQ-UHFFFAOYSA-N
Synonym dimethyl pyridin-2-ylboronate, pyridine-2-boronic acid dimethyl ester, dimethyl pyridineboronate, pyridine-2-boronic acid, dimethyl ester, dimethoxy pyridin-2-yl borane, 2-pyridylboronic acid, dimethyl ester, 2-pyridineboronic acid dimethyl ester, akos brn-0425, 2-dimethoxyboryl pyridine, acmc-209ca1
PubChem CID 2763164
IUPAC Name dimethoxy(pyridin-2-yl)borane
SMILES B(C1=CC=CC=N1)(OC)OC

Specifications

Quantity 250 mg
Formula Weight 150.97
Percent Purity 95%
Chemical Name or Material Pyridine-2-boronic acid dimethyl ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures