Learn More
Pyridine-2,6-dicarboxylic acid, 98%
CAS: 499-83-2 | C7H5NO4 | 167.12 g/mol
Supplier: Thermo Scientific Chemicals A1226322
Description
Pyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.
Solubility
Insoluble in water.
Notes
Incompatible with acids, bases and oxidizing agents.
Specifications
| Pyridine-2,6-dicarboxylic acid | |
| ∼245°C (decomposition) | |
| C7H5NO4 | |
| 100 g | |
| 2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid | |
| WJJMNDUMQPNECX-UHFFFAOYSA-N | |
| pyridine-2,6-dicarboxylic acid | |
| 10367 | |
| 167.12 |
| 499-83-2 | |
| 188°C (370°F) | |
| MFCD00006299 | |
| 131629 | |
| Insoluble in water. | |
| C1=CC(=NC(=C1)C(=O)O)C(=O)O | |
| 167.12 | |
| CHEBI:46837 | |
| 98% |
Chemical Identifiers
| 499-83-2 | |
| 167.12 | |
| WJJMNDUMQPNECX-UHFFFAOYSA-N | |
| 10367 | |
| pyridine-2,6-dicarboxylic acid |
| C7H5NO4 | |
| MFCD00006299 | |
| 2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid | |
| CHEBI:46837 | |
| C1=CC(=NC(=C1)C(=O)O)C(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-894-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only