missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2,3-dicarboxamide 98.0+%, TCI America™

Catalog No. p-7135012
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12155G 5 g
P121525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12155G Supplier TCI America Supplier No. P12155G
Only null left

Chemical Identifiers

CAS 4663-94-9
Molecular Formula C7H7N3O2
Molecular Weight (g/mol) 165.152
MDL Number MFCD00191488
InChI Key CJVCXRMYJNMDTP-UHFFFAOYSA-N
PubChem CID 578921
IUPAC Name pyridine-2,3-dicarboxamide
SMILES C1=CC(=C(N=C1)C(=O)N)C(=O)N

Specifications

Color White
Quantity 5 g
Formula Weight 165.15
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-2,3-dicarboxamide