Learn More
Corning™ PYREX™ VISTA™ Reusable Test Tubes with Beaded Rim
GREENER_CHOICE
Functionality, economy, and quality
Supplier:  Corning™ 7080025
 View more versions of this product
Catalog No. 07250119
Only null left
Description
Description
Functionality, economy, and quality
These test tubes are annealed, resistant to heat and chemically stable. Rims are fire-polished. The 25 mm diameter tubes do not have a marking spot.
Specifications
Chemical Identifiers
Beaded Rim
Borosilicate Glass
Non-sterile
No
Specifications
Beaded Rim
150 mm
Non-sterile
25 mm
55 mL
Borosilicate Glass
No
Test Tube
Product Suggestions
Product Suggestions