missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ VISTA™ Class B Volumetric Flasks with Polyethylene Snap-Cap
GREENER_CHOICE

Catalog No. 07770207
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
250 mL
500 mL
2000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
07770207 2000 mL
07770200 10 mL
07770201 25 mL
07770202 50 mL
07770204 250 mL
07770205 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 07770207 Supplier Corning™ Supplier No. 705812L

Capacity tolerances are twice those specified for Class A volumetric ware

These Class B volumetric flasks have capacity tolerances are twice those specified for Class A volumetric ware. The graduation line is sharp and permanent; large blue block letters make inscriptions easy to read. Snap caps are supplied with all sizes.

Specifications

Calibration To Contain
Closure Color Blue
Closure Material LDPE
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Neck Style Long
No. per Case 4/Cs.
No. per Pack 1/Pk.
Class B
Bottom Shape Flat
Closure Type Snap Cap
Show More Show Less