Learn More
PYREX™ Vacuum Distilling Adapter Tube
Converts flasks with standard taper necks to vacuum-type standard taper receivers
Supplier: PYREX™ 942024
Alert:
Because of variations in conditions, glassware cannot be guaranteed against breakage under vacuum or pressure. Adequate precautions should be taken to protect personnel who are doing such work.
Description
- Connecting tube with outer standard taper 24/40 joint at top, same size inner standard taper joint at bottom
- Side arm for rubber tubing is 0.39 in. (10mm) O.D. by 25mm long
Specifications
| Borosilicate Glass (3.3) | |
| 13.7 cm | |
| 24/40 Inner bottom, 24/40 outer top |
| 5.39 in. | |
| Converts flasks with necks to vacuum-type receivers | |
| Suction Tube Adapter |