missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Vacuum Desiccator Replacement Parts

Replacement cover sleeve rings for use with Pyrex desiccators.

$27.16 - $42.58

Specifications

Product Type Replacement Cover Sleeve Ring
No. per Pack 1/Pk.
No. per Case 1
View More Specs

Products 2
Catalog Number Mfr. No. Material For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Material For Use With (Application) Price Quantity & Availability  
086311A
View Documents Promotion Details
Corning™
3118RO
Borosilicate Glass For 2.2L desiccator
Case for $42.58
Only null left
 
086311C
View Documents Promotion Details
Corning™
3120RO
Plastic 3.8 to 7.5L Large Top Desiccator
Case for $27.16
Only null left
 
Description

Description

Replacement rings; reusable

  1. When ordering cover assembly, sleeve and ring must be purchased.
  2. For both 08-631A and 08-631B Desiccators.

Specifications

Specifications

Replacement Cover Sleeve Ring
1/Pk.
1
Videos
SDS
Documents

Documents