Learn More
Corning™ PYREX™ Squibb Separatory Funnel Without Valve And Stopper
Catalog No. 104443
Change view
Click to view available options
Capacity (Metric):
500 mL
Catalog No. Capacity (Metric)
104443 500 mL
1 options
Catalog No. 104443 Supplier Corning™ Supplier No. 6406500FO
Economy separatory funnel replacements
These PYREX economy Squibb separatory funnel replacements do not have a Polytetrafluorethylene (PTFE) valve assembly or polyethylene stopper. For replacement polyethylene Standard Taper stoppers, see No. 7624; for replacement PTFE locking stopcock assemblies, see No. 2116-LSO; for replacement tips, see No. 6406-GTO.
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass
For Use With (Application) For replacement polyethylene standard taper stoppers, for replacement PTFE locking stopcock assemblies
Quantity 4/Cs.
Stopcock PTFE
Stopper Number 27
Product Type Replacement Squibb Separatory Funnel