Learn More
Corning™ PYREX™ Solid Glass Pennyhead Standard Taper Stoppers
Description
Pennyhead standard taper stoppers are generally used in flasks and separatory funnels.
Specifications
Specifications
| Length (Metric) Ground Zone | 1.4 cm |
| Product Type | Pennyhead Stopper |
| For Use With (Equipment) | Flask and separatory funnel |
| Size | 9 |
| Material | Glass |