missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREX™ Reusable Test Tubes with Beaded Rim

Description
These tubes are made from special tubing to give the optimum wall thickness. They are well annealed, resistant to heat, and chemically stable. Rims are fire-polished.
Specifications
Specifications
| Design Type | Plain |
| Capacity (Metric) | 3 mL |
| Length (Metric) | 75 mm |
| Material | Borosilicate Glass |
| Diameter (Metric) | 10 mm |