Learn More
PYREX™ Regular Cyanide Distillation Apparatus
Catalog No. 0572316
Change view
Click to view available options
Type:
Cyanide distillation apparatus
Catalog No. Type
0572316 Cyanide distillation apparatus
1 options
Catalog No. 0572316 Supplier PYREX™ Supplier No. 3350C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
1L Two-neck boiling flask, cold finger condenser, condenser jacket, dispersion tube, distillation trap tube, inlet tube and Tygon tubing
For testing soluble and insoluble cyanides in water
Cyanide as hydrocyanic acid (HCN) is released from cyanide complexes by reflux distillation and absorbed in a scrubber containing sodium hydroxide solution.
  • Use for samples of 500mL
  • Reaction/distillation section of apparatus connects to absorption/scrubber section by Tygon™ tubing, reducing set-up time and eliminating a key source of glass breakage
  • Inlet tubes have funnel openings for easier acid addition
  • For replacement parts, see cat. nos. 0572316A - E
Compliance
Meets U.S. EPA SW-846 Methods 9010 and 9012 and ASTM™ Method D 2036.
Specifications
Certifications/Compliance US EPA SW-846 9010 and 9012, ASTM D2036
For Use With (Application) Testing soluble and insoluble cyanides in water
Includes 1L two neck boiling flask, cold finger condenser, condenser jacket, dispersion tube, distillation trap tube, inlet tube and tygon tubing
Type Cyanide distillation apparatus