Learn More
Corning™ PYREX™ PTFE Product Standard Straight Bore Stopcock Plug Assembly
GREENER_CHOICE
Replacement PTFE stopcock plug assemblies for straight bore PTFE stopcocks
$227.60 - $441.83
Spécifications
Material PTFE
No. per Pack 1/Pk.
No. per Case 1/Cs.
For Use With (Equipment) No. 7282 Pyrex straight bore PTFE stopcocks
voir les spécifications
Produits 3
Numéro de catalogue Numéro du manufacturier. Type Plug Bore Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Type Plug Bore Prix Quantity & Availability  
C76812
GREENER_CHOICE
Afficher les documents Détails de la promotion
Corning™
76812
Stopcock Plug Assembly, O-ring Size A 2 mm
caisse for $227.60
Il en reste null
C76816
GREENER_CHOICE
Afficher les documents Détails de la promotion
Corning™
76816
Stopcock Plug Assembly, O-ring Size B 6 mm
caisse for $441.83
Il en reste null
C76814
GREENER_CHOICE
Afficher les documents Détails de la promotion
Corning™
76814
Stopcock Plug Assembly, O-ring Size B 4 mm
caisse for $321.64
Il en reste null
Description
Description
These are replacement Polytetrafluorethylene (PTFE) Product Standard stopcock plug assemblies with various diameter bores for Cat. No. 7282 PYREX straight bore PTFE stopcocks. For more information on PTFE stopcocks, please check the Care of Corning PYREX™ PTFE Stopcocks under Technical Literature.
Order Info
Case of 1
Spécifications
Spécifications
PTFE
1/Pk.
1/Cs.
No. 7282 Pyrex straight bore PTFE stopcocks
Videos
FDS