Learn More
Corning™ PYREX™ Potter-Elvehjem Tissue Grinder with PTFE Pestle
Used in preparing small samples for pathological and biochemical studies
$368.40 - $553.44
Specifications
| Sterility | Non-sterile |
|---|---|
| Material | Borosilicate Glass |
| Product Type | Potter-Elvehjem Tissue Grinder |
| For Use With (Application) | Preparing small samples for pathological and biochemical studies |
| Includes | Case of 1 |
Alert:
If used with a motor-driven device, the attachment must contain some form of friction release clutch to prevent damage to the apparatus from pieces of connective tissue or muscle. The user should wear a protective glove of sufficient thickness to prevent injury in the event of failure of the glass components.
| Catalog Number | Mfr. No. | Capacity | Length (English) Tube | Tube Diameter (English) Outside | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity | Length (English) Tube | Tube Diameter (English) Outside | Price | Quantity | |||
|
09-552-26
|
Corning™
7725T1 |
0.03 oz., 1 mL | 3.9 in. | 0.5 in. |
Case for $390.98
|
|
|||
|
09-552-27
|
Corning™
7725T3 |
0.1 oz., 3 mL | 4.6 in. | 0.5 in. |
Case for $406.65
|
|
|||
|
09-552-31
|
Corning™
7725T45 |
1.5 oz., 45 mL | 8.7 in. | 1.2 in. |
Case for $553.44
|
|
|||
|
09-552-29
|
Corning™
7725T8 |
0.27 oz., 8 mL | 5.7 in. | 0.7 in. |
Case for $394.49
|
|
|||
|
09-552-25
|
Corning™
7725T5X |
0.01 oz., 0.5 mL | 3.2 in. | 0.5 in. |
Case for $439.34
|
|
|||
|
09-552-30
|
Corning™
7725T17 |
0.57 oz., 17 mL | 7.08 in. | 0.9 in. |
Case for $455.06
|
|
|||
|
09-552-28
|
Corning™
7725T5 |
0.16 oz., 5 mL | 5.5 in. | 0.6 in. |
Case for $368.40
|
|
|||
Description
Homogenization occurs as the sample and buffer are forced through the cylindrical portion of the mortar as the pestle is rotated downward. Mortar is unground and pestle contact surface is PTFE threaded onto a stainless steel shaft.
Caution: If used with a motor-driven device, the attachment must contain some form of friction release clutch to prevent damage to the apparatus. The user should wear a protective glove of sufficient thickness to prevent injury in the event of failure of the glass Âcomponents.
Order Info
Sold in a Case of 1.
Specifications
| Non-sterile | |
| Potter-Elvehjem Tissue Grinder | |
| Case of 1 | |
| PTFE |
| Borosilicate Glass | |
| Preparing small samples for pathological and biochemical studies | |
| Tooled mouth |