missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREX™ Polyethylene Snap Caps for Volumetric Flasks

Description
These blue caps provide an economical closure for certain volumetric flasks. Refer to volumetric flask listings to determine sizes and whether the flask is designed to take such a cap.
Specifications
Specifications
| Closure Color | Blue |
| Closure Material | HDPE |
| Color | Blue |
| For Use With (Equipment) | For volumetric flasks |
| No. per Case | 6/Cs. |
| Product Type | Cap, Snap |
| Disposable | Reusable |
| Closure Type | Snap Cap |
| Closure Size | 50 mm; Stopper No. 50 |