Learn More
Corning™ PYREX™ Polyethylene Snap Caps for Volumetric Flasks
Economical closure for some volumetric flasks
Supplier:  Corning™ 7666250
 View more versions of this product
Catalog No. C7666-250
Only null left
Explore available promotions
Description
Description
These blue caps provide an economical closure for certain volumetric flasks. Refer to volumetric flask listings to determine sizes and whether the flask is designed to take such a cap.
Specifications
Specifications
Blue
Blue
6/Cs.
Reusable
250
HDPE
For volumetric flasks
Cap, Snap
Snap Cap
Product Suggestions
Product Suggestions