missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Polyethylene Snap Caps for Volumetric Flasks
GREENER_CHOICE

Catalog No. C7666100
Change view
Click to view available options
Closure Size:
10
100
1000
200
2000
25
250
50 mm; Stopper No. 50
500
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
C7666100 100
C76661L 1000
C766625 25
C7666250 250
C7666500 500
C76662L 2000
C766650 50 mm; Stopper No. 50
C7666200 200
C766610 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. C7666100 Supplier Corning™ Supplier No. 7666100
Get Up to $2,500 in Products with Purchase
View all promotions

missing translation for 'validMainframeMsg'

Economical closure for some volumetric flasks

These blue caps provide an economical closure for certain volumetric flasks. Refer to volumetric flask listings to determine sizes and whether the flask is designed to take such a cap.

Specifications

Closure Color Blue
Closure Material HDPE
Color Blue
For Use With (Equipment) For volumetric flasks
No. per Case 6/Cs.
Product Type Cap, Snap
Disposable Reusable
Closure Type Snap Cap
Closure Size 100