Learn More
Corning™ PYREX™ Media Bottle Septa
For use with open-top PBT caps
$83.63 - $241.50
Specifications
| Color | Natural |
|---|---|
| No. per Case | 10/Cs. |
| No. per Pack | 10/Pk. |
| Quantity | 10 Cs. |
| Sterility | Non-sterile |
| Catalog Number | Mfr. No. | Material | For Use With (Equipment) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | For Use With (Equipment) | Price | Quantity & Availability | ||||
|
0641412
|
Corning™
139545SS |
Silicone Rubber | GL45 open top PBT screw cap |
Case of 10 Each for $241.50
|
|
||||
|
0641413
|
Corning™
139532SS |
Silicone Rubber | GL32 open top PBT screw cap |
Case of 10 Each for $91.09
|
|
||||
|
0641415
|
Corning™
139545TS |
PTFE-faced Silicone Rubber | GL45 open top PBT screw cap |
Case of 10 Each for $208.91
|
|
||||
|
0641416
|
Corning™
139532TS |
PTFE-faced Silicone Rubber | GL32 open top PBT screw cap |
Case of 10 Each for $130.00
|
|
||||
|
0641417
|
Corning™
139525TS |
PTFE-faced Silicone Rubber | GL25 open top PBT screw cap |
Case of 10 Each for $96.06
|
|
||||
|
0641414
|
Corning™
139525SS |
Silicone Rubber | GL25 open top PBT screw cap |
Case of 10 Each for $83.63
|
|
||||
Description
These are various sizes and types of screw caps for use with storage bottles.
Specifications
| Natural | |
| 10/Pk. | |
| Non-sterile |
| 10/Cs. | |
| 10 Cs. | |
| No Closure |