Learn More
Corning™ PYREX™ Reinforced Glass Bead Lifetime Red Cylinders, Single Metric Scale, TC
Calibrated To Contain
$103.63 - $2051.47
Specifications
Base Type Hexagonal
Calibration To Contain
Certifications/Compliance ASTM E1272, ASTM E133, ASTM E542, ASTM E694
Class Lifetime Red
Type Graduated Cylinder
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Qty  
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Qty  
08-551A
Accessories View Documents
Corning™
304610
10 mL 0.1 mL
Case of 24 Each for $1,866.67
Only null left
08-551B
Accessories View Documents
Corning™
304625
25 mL 0.2 mL
Each for $103.63
Case of 18 Each for $1,865.34
Only null left
08-551E
Accessories View Documents
Corning™
3046250
250 mL 2 mL
Each for $156.80
Case of 12 Each for $1,881.60
Only null left
08-551C
Accessories View Documents
Corning™
304650
50 mL 1 mL
Case of 18 Each for $2,051.47
Only null left
08-551D
Accessories View Documents
Corning™
3046100
100 mL 1 mL
Each for $132.59
Case of 12 Each for $1,591.08
Only null left
Description
Description
Calibrated 'To Contain.' The scale and figures are durable white enamel, which stand out clearly against the red color-band on these Lifetime Red cylinders. The 10 mL size has a funnel top. Bumper guards are supplied with 100 mL through 2000 mL inclusive. The 100 mL size is made in accordance with ASTM E-133 and E-1272 and is used in ASTM tests D-86 and D-216.
For the same cylinder, but with a built-in glass bumper guard, see Cat. No. 3046.
Compliance
100mL size in accordance with ASTM™ E 133 and E 1272; used in ASTM test D 86.
Specifications
Specifications
Hexagonal
ASTM E1272, ASTM E133, ASTM E542, ASTM E694
Graduated Cylinder
Round
Borosilicate Glass
Bumper Guard
Autoclavable
To Contain
Lifetime Red
White
Non-sterile
Accurate Measurements Delivering or Containing Liquids
Single