missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREX™ Large-Capacity Bottles with Tooled Neck
For distilled water and solutions
Supplier: Corning™ 159512
Description
For distilled water and solutions
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic
Order Info
Bottles take No. 12 size stoppers (not included).
Specifications
| 1536 oz., 45500 mL | |
| 1536 oz. | |
| Borosilicate Glass | |
| 1/Pk. | |
| No | |
| 584 mm | |
| 406 mm |
| 45,500 mL | |
| 15.9 in. | |
| 1/Cs. | |
| Non-sterile | |
| Cylindrical | |
| 22.9 in. |