missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ GL45 Screw Cap with Plug Seal

Catalog No. 064142C
Change view
Click to view available options
Closure Material:
PBT
Polypropylene
Closure Color:
Orange
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Material Closure Color
064142C PBT Red
064142A Polypropylene Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 064142C Supplier Corning™ Supplier No. 139545HTC

Nonsterile, reusable caps

These are various sizes and types of screw caps for use with storage bottles.

Specifications

Closure Color Red
Closure Material PBT
Color Red
For Use With (Equipment) Graduated Erlenmeyer flasks and media bottles
No. per Case 10/Cs.
No. per Pack 10/Pk.
Product Type Cap
Temperature Range (English) Up to 356°F
Temperature Range (Metric) Up to 180°C
Thread Size GL 45
Autoclavable Autoclavable
Disposable Reusable
Sterility Non-sterile
Closure Type High temperature
Show More Show Less