Learn More
Corning™ PYREX™ Glass Chromatographic Reagent Atomizer
GREENER_CHOICE
Used for spraying reagents onto thin layer chromatography plates
$718.11 - $764.11
Specifications
Material Pyrex Glass
Quantity 1/Cs.
Type Chromatographic Reagent Atomizer
For Use With (Application) For Spraying Reagents in Thin Layer Chromatography
Includes Full Length 24/40 joint
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Height (Metric) Price Quantity & Availability  
0572326A
GREENER_CHOICE
View Documents Promotion Details
Corning™
2153125
125 mL 18.8 cm
Case for $718.11
Only null left
0572326B
GREENER_CHOICE
View Documents Promotion Details
Corning™
2153250
250 mL 20.8 cm
Case for $764.11
Only null left
Description
Description
For use in spraying reagents in thin layer chromatography. With full length standard taper 24/40 joint between reservoir and spraying device.
Specifications
Specifications
Pyrex Glass
Chromatographic Reagent Atomizer
Full Length 24/40 joint
1/Cs.
For Spraying Reagents in Thin Layer Chromatography
24/40