missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREX™ Dounce Tissue Grinder with Two Pestles
Description
Includes
Unground glass tube (mortar) and two pestles
Supplied with two pestles to ensure dissociation of cells into fine particles with minimal damage to cell nuclei. Use the large clearance pestle for initial reduction of soft tissue. Complete the homogenization with the small clearance pestle. Particularly useful in enzyme studies where heat build-up must be avoided.
Specifications
Specifications
| Sterility | Non-sterile |
| Capacity (English) | 0.23 oz. |
| Capacity (Metric) | 7 mL |
| Material | Borosilicate Glass |
| Product Type | Dounce Tissue Grinder |
| For Use With (Application) | Ideal for Enzyme studies where heat build-up must be avoided, use large clearance pestle for initial reduction of soft tissue, use small clearance pestle to complete homogenization |
| Includes | Mortar tube, large pestle and small pestle |
| Length (English) Tube | 4.6 in. |
| Length (Metric) Tube | 117 mm |
| Tube Diameter (English) Outside | 0.59 in. |
| Show More |