Learn More
Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
Supplier:  Corning™ 9944516XX
 View more versions of this product
Catalog No. 149621B
Only null left
Description
Description
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
These one-use culture tubes are packaged for your convenience in easy-to-open, easy-to-use packs. The three small sizes come in a dispenser arrangement, and the larger sizes are bulk-packed. Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda-lime glass.
Specifications
Chemical Identifiers
Single-use
None
No
No
For tissue culture work and general bacteriological use
Bulk
Non-sterile
Culture Tube
Specifications
23 mL
Borosilicate Glass
For tissue culture work and general bacteriological use
Bulk
Non-sterile
16 mm
150 mm
Single-use
None
No
No
Culture Tube
Product Suggestions
Product Suggestions