Learn More
Corning™ PYREX™ Dispensing Burets with Straight Bore Product Standard PTFE Stopcock
For dispensing large volumes of liquids rapidly and accurately
Supplier: Corning™ 2094250
Description
Used when dispensing large volumes of liquids rapidly and accurately. Made from close tolerance, accurate bore tubing with uniform walls, these burets have a permanently colored scale. Tips are carefully drawn and ground to reduce chipping. Provided with a PTFE stopcock plug to reduce freezing and eliminate lubricant contamination. The smooth micro-finish of the stopcock barrel assures a precision fit and optimum leak-resistant performance.
Chemical Identifiers
| PTFE | |
| Dispensing Large Volumes of Liquids Rapidly and Accurately | |
| 1/Cs. | |
| PTFE |
| Glass | |
| 1/Pk. | |
| Straight Bore Product Standard PTFE Stopcock |
Specifications
| PTFE | |
| 33 mm | |
| 21.6 in. | |
| 1 mL | |
| 550 mm | |
| 1/Cs. | |
| 4 | |
| ±2 mL |
| 1.2 in. | |
| 250 mL | |
| Glass | |
| Dispensing Large Volumes of Liquids Rapidly and Accurately | |
| 1/Pk. | |
| Straight Bore Product Standard PTFE Stopcock | |
| PTFE |