Learn More
Corning™ PYREX™ Desiccator Replacement Bowls
Strong, easily cleaned, borosilicate glass bowls
Supplier:  Corning™ 3081200LO
 View more versions of this product
Catalog No. 08624416
Only null left
Description
Description
These PYREX™ desiccator bowls are replacements for No. 3081-150 2.4L desiccators. Warning: Do not heat or subject to pressure. Not for vacuum applications.
Order Info
  • The 2.4L sized bowl is a replacement for the Knob Top Desiccator 08-624-411 (Corning 3081-150) or the Small Top Desiccator (for use with 24/29 Stopcock) 08-624-420 (Corning 3121-150).
  • The 5.8L sized bowl is a replacement for either the Knob Top Desiccator 08-624-414 (Corning 3081-200) or the Large Top Desiccator (for use with 24/29 Stopcock) 08-624-422 (Corning 3121-200).
  • The 10.5L sized bowl is a replacement for either the Knob Top Desiccator 08-624-417 (Corning 3081-250) or the Large Top Desiccator (for use with 24/29 Stopcock) 08-624-424 (Corning 3121-250).
Specifications
Chemical Identifiers
Desiccator Replacement Bowl
1/Pk.
1
Reusable
Specifications
Desiccator Replacement Bowl
1
5.8L Large Knob Top Desiccator
7.95 in.
1/Pk.
Reusable
20.2 cm
223.5 mm
Product Suggestions
Product Suggestions