missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Cloning Cylinders
GREENER_CHOICE

Catalog No. p-176603
Change view
Click to view available options
Diameter (Metric):
6 mm
8 mm
10 mm
Height (Metric):
8 mm
10 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Height (Metric)
0955220 6 mm 8 mm
0955221 8 mm 8 mm
0955222 10 mm 10 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0955220 Supplier Corning™ Supplier No. 31666
Only null left

Create identical cells by surrounding a single cell or group of cells

Clone a single cell or group of cells by surrounding them with this glass cylinder. Dip the end of the cylinder into a sterile silicone grease before pressing to the bottom of a culture flask to create an ­isolated well.

Specifications

Diameter (Metric) 6 mm
Height (Metric) 8 mm
Sterility Non-sterile
Material Borosilicate Glass
Diameter (English) 0.23 in.
Product Type Cloning Cylinder
Height (English) 0.31 in.