Learn More
Corning™ PYREX™ Class B Lifetime Red™ Cylinders with Single Metric Scale, TC
Calibrated To Contain
Supplier:  Corning™ 3042500
 View more versions of this product
Catalog No. 08-552-1E
Only null left
Explore available promotions
Description
Description
Calibrated 'To Contain.' The scale and figures are durable white enamel, which stand out clearly against the red color-band on these Lifetime Red cylinders. The 10 mL size has a funnel top. Bumper guards are supplied with 100 mL through 2000 mL inclusive. The 100 mL size is made in accordance with ASTM E-133 and E-1272 and is used in ASTM tests D-86 and D-216.
For the same cylinder, but with a built-in glass bumper guard, see Cat. No. 3046.
Compliance
100mL size complies with ASTM™ E 133 and E 1272, and is used in ASTM test D 86.
Specifications
Specifications
Round
Graduated Cylinder
To Contain
White
Hexagonal
392 mm
1.9 in.
500 mL
Bumper Guard
5 mL
5 mL
±4 mL
15.3 in.
Non-sterile
ASTM E1272, ASTM E133, ASTM E542, ASTM E694
Lifetime Red
Borosilicate Glass
50 mm
Accurate Measurements Delivering or Containing Liquids
Single
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories