Learn More
Corning™ PYREX™ Class A Flasks Replacement Screw Caps
GREENER_CHOICE
Catalog No. 10207J
Change view
Click to view available options
No. per Case:
12/Cs.
Catalog No. No. per Case
10207J 12/Cs.
1 options
Catalog No. 10207J Supplier Corning™ Supplier No. 565018CO
Get Up to $2,500 in Products with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For 10-207 Series Class A flasks
These standard 18-410 G.P.I. thread screw caps are replacement caps for the No. 5650 PYREX(r) volumetric flasks. They have a durable cone-shaped polyethylene liner. This Poly-seal cap insures a leak-resistant closure and no contamination when working with most common reagents.
Specifications
Material Poly-seal
For Use With (Equipment) For Class A flasks
No. per Case 12/Cs.
No. per Pack 12/Pk.
Product Type Screw cap
Thread Size 18-410 GPI
Closure Type Screw Cap