Learn More
Corning™ PYREX™ Certified/Serialized Class A Cylinders with Metric Scale, TD
Calibrated To Deliver
$330.96 - $1018.13
Specifications
Base Type Hexagonal
Calibration To Deliver
Class Serialized/Certified Class A
Type Graduated Cylinder
Scale Color White
View More Specs
Includes: Certificate of Identification and Capacity traceable to standards provided by NIST™.
Products 8
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity  
08-553D
Accessories View Documents
Corning™
30621L
1 L 10 mL
Case for $639.12
Only null left
08-553G
Accessories View Documents
Corning™
306210
10 mL 0.1 mL
Case for $330.96
Only null left
08-553B
Accessories View Documents
Corning™
3062100
100 mL 1 mL
Case for $396.11
Only null left
08-553E
Accessories View Documents
Corning™
306225
25 mL 0.2 mL
Case for $355.17
Only null left
08-553A
Accessories View Documents
Corning™
306250
50 mL 1 mL
Case for $372.03
Only null left
08-553C
Accessories View Documents
Corning™
3062500
500 mL 5 mL
Case for $537.47
Only null left
08-553H
Accessories View Documents
Corning™
30622L
2000 mL 20 mL
Case for $1,018.13
Only null left
08-553F
Accessories View Documents
Corning™
3062250
250 mL 2 mL
Case for $459.15
Only null left
Description
Description
  • Calibrated “To Deliver” in accordance with ASTM E-1272 and ASTM E-542.
  • Each cylinder is individually serialized and supplied with a Certificate of Identification and Capacity, traceable to NIST standards.
  • Permanent white graduations are easy to read.
  • Bumper guards are supplied with 25 mL through 2000 mL inclusive.
  • The 10 mL size has a funnel top.
  • Tested and calibrated in an ISO/IEC 17025 certified laboratory.
Compliance
Class A tolerances in accordance with ASTM™ E 1272 and E 542.
Specifications
Specifications
Hexagonal
Serialized/Certified Class A
White
Non-sterile
Case of 1
Autoclavable
To Deliver
Graduated Cylinder
Round
Borosilicate Glass
Single
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'