Learn More
Corning™ PYREX™ Buchner Funnels with Fine Porosity Fritted Disc
Useful where filter paper would be attacked by material being filtered
Supplier:  Corning™ 360602C
 Afficher plus de versions de ce produit
Numéro de catalogue. 20704A
Il en reste null
Description
Description
Useful where filter paper would be attacked by material being filtered
These funnels are especially useful where filter paper would be attacked by the material being filtered. The disc should not be subjected to pressure differential in excess of 1 atmosphere. The top edge is beaded.
Spécifications
Identifiants chimiques
Borosilicate Glass
Useful where filter paper would be attacked by material being filtered
Buchner Funnel
ASTM E128
Fritted discs
Spécifications
2 mL
Borosilicate Glass
ASTM E128
10 mm
31 mm
40 to 60 μm
7 mm
70 mm
Useful where filter paper would be attacked by material being filtered
Fritted discs
Coarse
Buchner Funnel
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Accessoires