missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1086375-50-9
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD09414725
InChI Key SBDWLDBGGVPVAR-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyrimidine-5-carboxylic acid, pyrazolo 1?5-a pyrimidine-5-carboxylic acid, 8-hydropyrazolo 1,5-a pyrimidine-5-carboxylic acid
PubChem CID 28875802
IUPAC Name pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
SMILES OC(=O)C1=NC2=CC=NN2C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC432720010
View Documents Promotion Details
N/A 1g Glass bottle N/A
Only null left
 
AC432720050
View Documents Promotion Details
N/A 5g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1086375-50-9
163.14
SBDWLDBGGVPVAR-UHFFFAOYSA-N
28875802
OC(=O)C1=NC2=CC=NN2C=C1
C7H5N3O2
MFCD09414725
pyrazolo 1,5-a pyrimidine-5-carboxylic acid, pyrazolo 1?5-a pyrimidine-5-carboxylic acid, 8-hydropyrazolo 1,5-a pyrimidine-5-carboxylic acid
pyrazolo[1,5-a]pyrimidine-5-carboxylic acid

Specifications

1086375-50-9
96% min. (HPLC)
MFCD09414725
SBDWLDBGGVPVAR-UHFFFAOYSA-N
pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
28875802
97%
Authentic
C7H5N3O2
pyrazolo 1,5-a pyrimidine-5-carboxylic acid, pyrazolo 1?5-a pyrimidine-5-carboxylic acid, 8-hydropyrazolo 1,5-a pyrimidine-5-carboxylic acid
OC(=O)C1=NC2=CC=NN2C=C1
163.14
163.14
Pyrazolo[1, 5-a]pyrimidine-5-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Take off con

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only