missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid, 97%

Catalog No. AC432720050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432720050 5 g Glass bottle
AC432720010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432720050 Supplier Thermo Scientific Chemicals Supplier No. 432720050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1086375-50-9 | C7H5N3O2 | 163.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1086375-50-9
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD09414725
InChI Key SBDWLDBGGVPVAR-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyrimidine-5-carboxylic acid, pyrazolo 1?5-a pyrimidine-5-carboxylic acid, 8-hydropyrazolo 1,5-a pyrimidine-5-carboxylic acid
PubChem CID 28875802
IUPAC Name pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
SMILES OC(=O)C1=NC2=CC=NN2C=C1

Specifications

Melting Point 222°C to 225°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 163.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Pyrazolo[1, 5-a]pyrimidine-5-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Take off con

RUO – Research Use Only