missing translation for 'onlineSavingsMsg'
Learn More

Pyrazole-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. P19785G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19785G 5 g
P19781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19785G Supplier TCI America Supplier No. P19785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37718-11-9
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.088
MDL Number MFCD00077436
InChI Key IMBBXSASDSZJSX-UHFFFAOYSA-N
Synonym 4-pyrazolecarboxylic acid, 4-carboxypyrazole, pyrazole-4-carboxylic acid, unii-4xjz84gv8e, 1h-pyrazole-4-carboxylicacid, 4-carboxy-1h-pyrazole, 4xjz84gv8e, pyrazole-4-carboxylic, pubchem13293, pubchem22749
PubChem CID 3015937
IUPAC Name 1H-pyrazole-4-carboxylic acid
SMILES C1=C(C=NN1)C(=O)O

Specifications

Melting Point 275°C
Color White
Quantity 5 g
Formula Weight 112.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrazole-4-carboxylic Acid