missing translation for 'onlineSavingsMsg'
Learn More

Pyrazole-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7135294
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
P18621G 1 g
P18625G 5 g
2 options

Catalog No. P18621G

Supplier: TCI America P18621G

Only null left

Chemical Identifiers

CAS 1621-91-6
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00077436
InChI Key KOPFEFZSAMLEHK-UHFFFAOYSA-N
Synonym 1h-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 2h-pyrazole-3-carboxylic acid, 5-pyrazolecarboxylic acid, 3-pyrazolecarboxylic acid, 3-carboxy-1h-pyrazole, pyrazolic acid, pyrazolecarboxylic, 3-carboxypyrazole, 2h-pyrazole-3-carboxylicacid
PubChem CID 574310
IUPAC Name 1H-pyrazole-5-carboxylic acid
SMILES OC(=O)C1=CC=NN1

Specifications

Melting Point 213°C
Color White-Yellow
Quantity 1 g
Formula Weight 112.09
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pyrazole-3-carboxylic Acid