missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinecarbonitrile, 99%

Catalog No. AC436920100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436920100 10 g Glass bottle
AC436920500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436920100 Supplier Thermo Scientific Chemicals Supplier No. 436920100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19847-12-2 | C5H3N3 | 105.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19847-12-2
Molecular Formula C5H3N3
Molecular Weight (g/mol) 105.10
MDL Number MFCD00049361
InChI Key PMSVVUSIPKHUMT-UHFFFAOYSA-N
Synonym pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine
PubChem CID 73172
ChEBI CHEBI:3982
IUPAC Name pyrazine-2-carbonitrile
SMILES N#CC1=CN=CC=N1

Specifications

Melting Point 20°C
Color Colorless to Yellow
Density 1.1740g/mL
Boiling Point 90°C to 93°C (15.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5330 to 1.5350 (20°C, 589nm)
Quantity 10 g
Specific Gravity 1.174
Solubility Information Solubility in water: soluble
Formula Weight 105.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Pyrazinecarbonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 243-369-5

RUO – Research Use Only