missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrazine 98.0+%, TCI America™
$37.29 - $447.38
Chemical Identifiers
| CAS | 290-37-9 |
|---|---|
| Molecular Formula | C4H4N2 |
| Molecular Weight (g/mol) | 80.09 |
| MDL Number | MFCD00006122 |
| InChI Key | KYQCOXFCLRTKLS-UHFFFAOYSA-N |
| Synonym | 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; |
| PubChem CID | 9261 |
| ChEBI | CHEBI:30953 |
| IUPAC Name | pyrazine |
| SMILES | C1=CN=CC=N1 |
Chemical Identifiers
| 290-37-9 | |
| 80.09 | |
| KYQCOXFCLRTKLS-UHFFFAOYSA-N | |
| 9261 | |
| pyrazine |
| C4H4N2 | |
| MFCD00006122 | |
| 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; | |
| CHEBI:30953 | |
| C1=CN=CC=N1 |
Specifications
| 290-37-9 | |
| White | |
| C4H4N2 | |
| 25 g | |
| 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; | |
| C1=CN=CC=N1 | |
| 80.09 | |
| CHEBI:30953 | |
| ≥98.0% (GC) | |
| Pyrazine |
| 54°C | |
| 118°C | |
| MFCD00006122 | |
| 1325 | |
| KYQCOXFCLRTKLS-UHFFFAOYSA-N | |
| pyrazine | |
| 9261 | |
| 80.09 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3827
RTECSNumber : UQ2015000
TSCA : Yes