Learn More
Pyrazine, 99+%
CAS: 290-37-9 | C4H4N2 | 80.09 g/mol
$89.89 - $421.84
Chemical Identifiers
| CAS | 290-37-9 |
|---|---|
| Molecular Formula | C4H4N2 |
| Molecular Weight (g/mol) | 80.09 |
| InChI Key | KYQCOXFCLRTKLS-UHFFFAOYSA-N |
| Synonym | 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; |
| PubChem CID | 9261 |
| ChEBI | CHEBI:30953 |
| IUPAC Name | pyrazine |
| SMILES | C1=CN=CC=N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC131700100
|
Thermo Scientific Chemicals
131700100 |
10 g | Glass bottle |
Each for $89.89
|
|
||||
|
AC131700250
|
Thermo Scientific Chemicals
131700250 |
25 g | Glass bottle |
Each for $144.24
|
|
||||
|
AC131701000
|
Thermo Scientific Chemicals
131701000 |
100 g | Glass bottle |
Each for $421.84
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 290-37-9 | |
| 80.09 | |
| 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; | |
| CHEBI:30953 | |
| C1=CN=CC=N1 |
| C4H4N2 | |
| KYQCOXFCLRTKLS-UHFFFAOYSA-N | |
| 9261 | |
| pyrazine |
Specifications
| 290-37-9 | |
| White | |
| 55°C | |
| 99+% | |
| C4H4N2 | |
| 23,91 | |
| 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; | |
| KYQCOXFCLRTKLS-UHFFFAOYSA-N | |
| pyrazine | |
| 9261 | |
| 80.09 | |
| Crystals |
| 52.0°C to 56.0°C | |
| 115.0°C to 116.0°C | |
| Authentic | |
| Glass bottle | |
| 10 g | |
| 15,8068 | |
| Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol and ether | |
| C1=CN=CC=N1 | |
| 80.09 | |
| CHEBI:30953 | |
| 99+% | |
| Pyrazine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medi
GHS Signal Word: Warning
EINECSNumber : 206-027-6
RUO – Research Use Only