missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine, 98%

CAS: 290-37-9 | C4H4N2 | 80.09 g/mol

$74.51 - $187.12

Chemical Identifiers

CAS 290-37-9
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006122
InChI Key KYQCOXFCLRTKLS-UHFFFAOYSA-N
Synonym 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine;
PubChem CID 9261
ChEBI CHEBI:30953
IUPAC Name pyrazine
SMILES C1=CN=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1632609
View Documents
Thermo Scientific Chemicals
01632609
10 g
Each for $74.51
Only null left
 
AA1632618
View Documents
Thermo Scientific Chemicals
01632618
50 g
Each for $187.12
Only null left
 
Description

Description

Pyrazine is used as a flavoring agent. It serves as a component of some herbs in traditional Chinese medicine. It is employed as pharmaceutical intermediates, perfume intermediates and involved in the preparation of other chemicals. In addition to this, it is used in the production of pyrazine 1-oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

290-37-9
80.09
KYQCOXFCLRTKLS-UHFFFAOYSA-N
9261
pyrazine
C4H4N2
MFCD00006122
1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine;
CHEBI:30953
C1=CN=CC=N1

Specifications

290-37-9
White
113°C to 116°C
Strong
1.5235
10 g
103905
14,7957
Soluble in water,alcohol,and ether.
C1=CN=CC=N1
80.09
CHEBI:30953
98%
Pyrazine
50°C to 56°C
1.031 g/mL
55°C (131°F)
C4H4N2
MFCD00006122
UN1325
Hygroscopic
1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine;
KYQCOXFCLRTKLS-UHFFFAOYSA-N
pyrazine
9261
80.09
Crystalline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 206-027-6

RTECSNumber : UQ2015000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only