missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2-carbonitrile, 97+%

Catalog No. p-7081142
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1097414 25 g
AAA1097422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1097414 Supplier Thermo Scientific Chemicals Supplier No. A1097414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19847-12-2 | C5H3N3 | 105.10 g/mol

Pyrazine-2-carbonitrile is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrazine-2-carbonitrile is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 19847-12-2
Molecular Formula C5H3N3
Molecular Weight (g/mol) 105.10
MDL Number MFCD00049361
InChI Key PMSVVUSIPKHUMT-UHFFFAOYSA-N
Synonym pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine
PubChem CID 73172
ChEBI CHEBI:3982
IUPAC Name pyrazine-2-carbonitrile
SMILES N#CC1=CN=CC=N1

Specifications

Melting Point 18°C to 20°C
Density 1.174
Boiling Point 90°C to 93°C (15 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.536
Quantity 25 g
UN Number UN3276
Beilstein 110008
Solubility Information Slightly soluble in water.
Formula Weight 105.1
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Pyrazine-2-carbonitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H StatementH331-H302-H312-H315-H319-H335 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 243-369-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only