missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2,3-dicarboxylic acid, 98%

Catalog No. AAB2210806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2210806 5 g
AAB2210814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2210806 Supplier Thermo Scientific Chemicals Supplier No. B2210806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 89-01-0 | C6H4N2O4 | 168.108 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-01-0
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.108
MDL Number MFCD00006131
InChI Key ZUCRGHABDDWQPY-UHFFFAOYSA-N
Synonym 2,3-pyrazinedicarboxylic acid, pyrazine-2,3-dicarboxylate, 2,3-dicarboxypyrazine, 2,3-pyrazine dicarboxylic acid, unii-qnn35wof29, 2,3-pyrazinedicarboxylicacid, qnn35wof29, 2,3-pyrazinedcarboxylicacid, pubchem8610, acmc-209qye
PubChem CID 66628
IUPAC Name pyrazine-2,3-dicarboxylic acid
SMILES C1=CN=C(C(=N1)C(=O)O)C(=O)O

Specifications

Melting Point ∼188°C (decomposition)
Quantity 5 g
Beilstein 147982
Merck Index 14,7958
Formula Weight 168.11
Percent Purity 98%
Chemical Name or Material Pyrazine-2,3-dicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-875-3
RTECSNumber UQ2500000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only