missing translation for 'onlineSavingsMsg'
Learn More

Pyrantel Impurity D Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002105
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002105 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002105 Supplier Merck Emd Millipore Supplier No. PHR208450MG
Only null left

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 4271-96-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C6H12N2
Synonym 1,2-Dimethyl-1,4,5,6-tetrahydropyrimidine
Type Certified Reference Material
Quantity 50 mg
Formula Weight 112.17
Grade Certified Reference Material