Learn More
Pyramex™ Ztek™ Safety Glasses: Reading Lenses
Description
- Lightweight, wraparound frame/lens combination provides superior side protection, without the distraction of seams or bends
- Soft, rubber temple tips provide comfort without slipping
- Coated, scratch-resistant lenses provide 99% UV protection
Compliance
Meet ANSI Z87.1 requirements and CSA-Z94.3 standards
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Certifications/Compliance | Meet ANSI Z87.1 requirements and CSA Z94.3 standards |
| Description | +2.00 Reading lens |
| Lens Tint | Clear |
| Frame Color | Clear |
| Magnification Power | +2.00 |
| Lens Material | Polycarbonate |