Learn More
Pyramex™ Ztek™ Safety Glasses
Catalog No. 296352978
Change view
Click to view available options
Frame Color:
Amber
Black
Blue
Clear
Clear Gray
White
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Lens Tint:
Amber
Blue Mirror
Clear
Coffee
Gold Mirror
Gray
Indoor/Outdoor Mirror
Infinity Blue
Silver Mirror
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
296352978 Clear Anti-fog, Scratch-resistant Clear
19151315 Black Scratch-resistant Coffee
19151318 Blue Scratch-resistant Infinity Blue
19151319 Black Scratch-resistant Silver Mirror
19151317 Amber Scratch-resistant Amber
19151320 Black Scratch-resistant Blue Mirror
19151322 Black Scratch-resistant Gold Mirror
19817359 Clear Scratch-resistant Indoor/Outdoor Mirror
19151316 Black Scratch-resistant Gray
19817358 White Scratch-resistant Clear
19817420 Clear Gray Anti-fog, Scratch-resistant Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 296352978 Supplier Pyramex™ Supplier No. S2510ST
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Wraparound single lens with panoramic view
  • Lightweight, wraparound frame/lens combination provides superior side protection, without the distraction of seams or bends
  • Soft, rubber temple tips provide comfort without slipping
  • Coated, scratch-resistant lenses provide 99% UV protection
Compliance
Meet ANSI Z87.1 requirements and CSA-Z94.3 standards
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance Meet ANSI Z87.1 requirements and CSA Z94.3 standards
Lens Tint Clear
Product Type Safety Glasses
Frame Color Clear
Temple Style Rubber coated tips