Learn More
Pyramex™ Ztek™ Safety Glasses
Wraparound single lens with panoramic view
Supplier: Pyramex™ S2520S
Description
- Lightweight, wraparound frame/lens combination provides superior side protection, without the distraction of seams or bends
- Soft, rubber temple tips provide comfort without slipping
- Coated, scratch-resistant lenses provide 99% UV protection
Compliance
Meet ANSI Z87.1 requirements and CSA-Z94.3 standards
Specifications
| Scratch-resistant | |
| Gray | |
| Black |
| Meet ANSI Z87.1 requirements and CSA Z94.3 standards | |
| Safety Glasses | |
| Rubber coated tips |